Thermo Scientific™ Pierce™ Mouse Soluble ICAM-1 (CD54) ELISA Kits

Measure levels of mouse soluble ICAM-1 (CD54) in mouse serum, plasma (EDTA) and cell culture supernatant samples using 96-well plates.

Overview
Brand: Thermo Scientific™ Pierce™

Manufacturer Part Number: EMICAM1

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.45000kg Transport: UN number : 3261 Chem class : 1a Pack group : III



Product Code. 10381104

Quantity Price
1 £ 407.0 / 1 set
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Precision Intra-assay CV <10%; inter assay CV <10%
Sensitivity <5pg/mL
Formulation Complete ELISA kit with coated 96-well plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Kit Contents 30X Wash Buffer, 50mL
Sufficient For 96 microplate wells
Assay Range 25.6 to 1000pg/mL
Specificity No significant cross reaction with rat or human sICAM 1
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Mouse soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54)
Sample Size 50µL of 100-fold diluted samples per well
Description Mouse Soluble ICAM-1 (CD54) ELISA Kit
Total Assay Time 3.5 hr.
Quantity 1 plate kit

These mouse sICAM-1 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54) in samples at 50μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Mouse soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 25.6 to 1000ng/mL

  • Sensitivity: < 5ng/mL

  • Precision: Intra-assay CV < 10%; inter assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL of 100 fold diluted samples per well

  • Specificity: No significant cross reaction with rat or human sICAM 1

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Documentation

Technical documents