Learn More
R&D Systems™ Mouse/Rat Total Cytochrome c DuoSet IC, Short Format, R&D Systems™
Mouse/Rat Total Cytochrome c DuoSet IC ELISA, Short Format from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DYC897-5
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13434845
  • £827.61 / Pack of 5
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
The Mouse/Rat Total Cytochrome c DuoSet IC ELISA, Short Format is a Solid Phase Sandwich ELISA that quantifies mouse, rat Cytochrome c.
Specifications
Specifications
ELISA
5 x 96-well plate
1 kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
ELISA kit
Mouse, rat