missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse/Rat IL-17A ELISPOT ELISA Ready-SET-Go!™ Kit

This Mouse IL-17A ELISPOT Ready-SET-Go! reagent set contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IL-17A-secreting cells. This ELISPOT reagent set is pre-titrated for optimal spot development.

Brand:  Invitrogen™ 88-7370-21

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15571127

  • £152.25 / Pack of 2


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISPOT Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 uM).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

Description

Description

The complete, ready-to-use set includes all the reagents needed to perform ELISPOT assays, including: capture and detection antibody, coating buffer powder, assay diluent and detection enzyme. It is supplied with a certificate of anaylsis.

Specifications

Specifications

ELISPOT
Mouse
IL-17A
2 x 96 tests
2° to 8°C
Interleukin-17A
Videos

For Research Use Only