missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat IGF-I Quantikine ELISA Kit, R&D Systems™

Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SMG100

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13432816

  • £3167.85 / Pack of 6


Estimated Shipment date
from Supplier 03-04-2020
Add to basket

Description

Description

The Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat IGF-I/IGF-1 in cell culture supernates (25 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications

Specifications

31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
24482
8.4pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
1 Pack
Tissue Homogenates
HRP
ELISA
IGF-I Quantikine ELISA Kit
Cell Culture Supernates (25uL),Tissue Homogenates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Mouse, Rat
Mouse, Rat
Videos
Documents