missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat CCL2/JE/MCP-1 Quantikine ELISA Kit, R&D Systems™

A 4.5 hour solid phase ELISA designed to measure MCP-1 in mouse or rat cell culture supernates and serum. It contains E. coli-expressed mouse MCP-1 and antibodies raised against the recombinant factor.

Brand:  R&D Systems™ SMJE00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462816

  • £1852.45 / Pack of 6


Estimated Shipment
01-03-2019
Add to basket

Description

Description

MCP-1, also known as monocyte chemotactic and activating factor (MCAF), was initially purified based on its ability to chemoattract monocytes. Subsequent to its cloning and sequencing, it became evident that this protein is also identical to the product of the human JE gene. The JE gene, originally identified in mouse fibroblasts, is a platelet-derived growth factor (PDGF)-inducible gene.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (25μL), Serum (25μL)
Sensitivity: 2pg/mL
Specificity: Natural and recombinant mouse and rat MCP-1
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

15.60 - 1000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
20296 (Mouse); 24770 (Rat)
2pg/mL
Cell Culture Supernates (25μL), Serum (25μL)
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
1 Pk.
Cell Culture Supernates (25uL), Serum (25uL)
Mouse, Rat
Videos