Learn More
Invitrogen™ PRDC (GREM2) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMGREM2
Code : NEW
Additional Details : Weight : 0.00400kg
Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Mouse PRDC (GREM2) ELISA quantitates Ms PDRC in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms PDRC. Principle of the method The Mouse PDRC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation.Specifications
O88273 | |
3 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
23893 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Bone Research | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
3.28-800 pg/mL | |
3pg/mL to 800pg/mL | |
Biotin | |
Other Proteins | |
This ELISA pair antibody detects mouse PRDC. Other species not determined. | |
Mouse | |
Colorimetric Microplate Reader | |
Grem2 | |
<10% | |
HRP | |
RUO | |
Plasma, 33 μL; Serum, 33 μL; Supernatant, 100 μL | |
Gremlin2, Prdc | |
1 hr. 20 min. |