missing translation for 'onlineSavingsMsg'
Learn More

Mouse Phospho-Tie-2 DuoSet IC, R&D Systems™

Mouse Phospho-Tie-2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  RD Systems (Bio-Techne) DYC2816-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454525

  • £441.00 / Pack of 2


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

The Mouse Phospho-Tie-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated mouse Tie-2.
Specifications

Specifications

ELISA
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
Mouse
1 Kit
21687
2 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Videos
Documents