missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIP-3 alpha (CCL20) Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMCCL20

Code : NEW

Additional Details : Weight : 0.33000kg

Product Code. 15405233

  • £398.00 / 1 set


Estimated Shipment date
from Supplier 29-06-2020
Add to basket

Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

Macrophage Inflammatory Protein-3 alpha (MIP-3 alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3 alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
Specifications

Specifications

O89093
HRP
3pg/mL to 1500pg/mL
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
100 μL per sample after dilution
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, CD30L, CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL- 4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
96 assays
4 hr. 45 min.
Solid-phase sandwich ELISA
Inflammation
CKb4, LARC, MIP-3A, MIP-3[a], MIP3A, ST38, Scya20, exodus-1
<10%
Mouse
Colorimetric Microplate Reader
2.06-1500 pg/mL
Biotin
20297
ELISA Kit
3 pg/mL
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL
2°C to 8°C
1 hr. 20 min.
Mouse
Cytokines & Receptors
Ccl20
<12%
96 Tests
Supernatant
Videos
Documents