missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIP-1 gamma (CCL9) Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMCCL9

Code : NEW

Additional Details : Weight : 0.33000kg

Product Code. 15495233

  • £398.00 / 1 set


Estimated Shipment date
from Supplier 02-06-2020
Add to basket

Description

Description

Monokine with inflammatory, pyrogenic and chemokinetic properties. Circulates at high concentrations in the blood of healthy animals. Binding to a high-affinity receptor activates calcium release in neutrophils. It also inhibits colony formation of bone marrow myeloid immature progenitors.
Specifications

Specifications

P51670
HRP
1pg/mL to 250pg/mL
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
100 μL per sample after dilution
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 delta, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
96 assays
4 hr. 45 min.
Solid-phase sandwich ELISA
Inflammation
RP23-320E6.3, CCF18, MRP-2, Scya10, Scya9
<10%
Mouse
Colorimetric Microplate Reader
1.02-250 pg/mL
Biotin
20308
ELISA Kit
1 pg/mL
Plasma, 2.5 μL|Serum, 2.5 μL|Supernatant, 100 μL
2°C to 8°C
1 hr. 20 min.
Mouse
Chemokine & Receptors
CCF18,MRP-2,Scya10,Scya9
<12%
96 Tests
Supernatant
Videos
Documents