Thermo Scientific™ Pierce™ Mouse MCP-1 ELISA Kits

Measure levels of mouse monocyte chemoattractant protein-1 (MCP1) in mouse serum, plasma and cell culture supernatant samples using 96-well plates.

Overview
Brand: Thermo Scientific™ Pierce™

Manufacturer Part Number: EMMCP12

MOUSE MCP-1 ELISA

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10107593

Quantity Price
1 £ 443.0 / 1 set
Estimated Shipment date
from Supplier 03-02-2017
Add to basket


Description and Specification

Specification

Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <4pg/mL
Formulation Complete ELISA kit with coated 96-well plates
Measurement Absorbance at 450nm minus absorbance at 550nm
Kit Contents 30X Wash Buffer, 2 x 50mL
Assay Range 16 to 1000pg/mL
Sufficient For 2 x 96 microplate wells
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Specificity No significant cross-reaction with mouse G-CSF, GM-CSF, IFNg, IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MIP-1a, RANTES or TNFa, or human or rat MCP-1
Description Mouse MCP-1 ELISA Kit
Sample Size 50.0µL
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant mouse MCP-1 (monocyte chemoattractant protein-1)
Total Assay Time 3 hr.
Quantity 2 plate kit

These mouse MCP-1 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of monocyte chemoattractant protein-1 (MCP-1) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse MCP-1 (monocyte chemoattractant protein-1)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 16 to 1000pg/mL

  • Sensitivity: < 4pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse G-CSF, GM-CSF, IFNγ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MIP-1α, RANTES or TNFα, or human or rat MCP-1

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates.

Documentation

Technical documents