missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Stem Cell Factor (SCF), Recombinant Protein

Mouse Recombinant Protein

Brand:  Invitrogen™ 14-8341-62

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15557526

  • £88.03 / 5µg


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Description

Description

Mouse Stem Cell Factor (SCF) is a hematopoietic growth factor and ligand for the KIT tyrosine kinase receptor, which exerts its activity in the early stages of hematopoiesis. Mouse SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures.
Specifications

Specifications

M59915
Sterile liquid; phosphate buffered saline, pH 7.2, 1.0% BSA. 0.22μm filtered.
Stem Cell Factor (SCF)
≥95%
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
The ED50 of this protein, as measured by MC/9 cell proliferation assay, is less than or equal to 65ng/mL. This corresponds to a specific activity of greater than or equal to 1.5 x 104 Units/mg.
Unlabeled
Yes
Cytokine Bioassays
The protein is not methionylated at the N-terminal and has a predicted molecular mass of 18,298. The DTT reduced protein migrates as an 18kDa polypeptide on SDS-PAGE. The non-reduced protein migrates as a 16kDa polypeptide.
SDS-PAGE
5μg
Store at less than or equal to -70°C.
E. coli expressed amino acids Lys 26-Ala 189 of mature mouse SCF accession # M59915
kit-ligand, KL, steel factor
Recombinant Protein
Mouse
Videos