missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-6 Quantikine ELISA Kit, R&D Systems™

Mouse IL-6 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SM6000B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462806

  • £2266.74 / Pack of 6


Estimated Shipment date
from Supplier 30-03-2020
Add to basket

Description

Description

The Mouse IL-6 Quantikine ELISA Kit quantifies mouse IL-6 in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul).
Specifications

Specifications

1 Pack
Mouse
HRP
ELISA
Solid Phase Sandwich ELISA
1.8pg/mL
Natural and recombinant mouse IL-6
Sufficient materials to run ELISAs on at least six 96-well plates
Serum
7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
16193
6 Plates
Cell Culture Supernates (50uL),Serum (50uL),Heparin Plasma (50uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
B cell stimulatory factor-2, B-cell differentiation factor, BSF2, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Videos
Documents