missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-6 Platinum ELISA Kit

Specifically engineered for accurate and precise measurement of IL-6 protein levels from samples including serum, plasma, and supernatants from cell cultures

Brand:  Invitrogen™ BMS603-2TEN

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15532007

  • £1565.85 / Pack of 10


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-6
Biotin-Conjugate anti-mouse IL-6 monoclonal antibody
Streptavidin-HRP
Mouse IL-6 Standard lyophilized, 4ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Mouse IL-6 Platinum ELISA contains the necessary reagents, from pre-coated plates to standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
Mouse
Interferon-beta 2, B-cell stimulatory factor 2,HGF
ELISA
6.5pg/mL
50μL
IL-6
Videos