missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-6 Mouse ELISA Kit
Specifically engineered for accurate and precise measurement of IL-6 protein levels from samples including serum, plasma, and supernatants from cell cultures
£451.00 - £2400.00
Specifications
Accession Number | P08505 |
---|---|
Product Type | ELISA Kit |
Sample Type | Plasma, Serum, Supernatant |
Target Species | Mouse |
For Use With (Application) | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15522007
|
Invitrogen™
BMS603-2 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
15542007
|
Invitrogen™
BMS603-2TWO |
2 x 96 Tests |
£580.00
Pack of 2 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15532007
|
Invitrogen™
BMS603-2TEN |
10 x 96 Tests |
£2400.00
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
Mouse IL-6 Platinum ELISA contains the necessary reagents, from pre-coated plates to standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
P08505 | |
Plasma, Serum, Supernatant | |
ELISA | |
16193 | |
5% | |
RUO | |
2°C to 8°C | |
Mouse | |
3 hr. 30 min. |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
8.9% | |
HRP | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
Interferon-beta 2, B-cell stimulatory factor 2,HGF | |
1 hr. 20 min. |
For Research Use Only