missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-6 ELISA Kits

Measure levels of mouse interleukin-6 in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ EM2IL6

Code : PN

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10751175

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

These mouse IL-6 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The antibody-pair kit consists of the matched IL-6 antibody pairs and IL-6 standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse interleukin-6 (IL-6)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 51 to 2000pg/mL

  • Sensitivity: < 7pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse GM-CSF, IFNγ, IL-1α, IL-2, IL-3, IL-4, IL-5, IL-7, IL-9, IL-10, IL-12, or human IL-6. This kit has been shown to have a high level of cross-reactivity with recombinant rat IL-6

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the antibody-pair kit.

Includes:The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates.

Specifications

Specifications

51 to 2000pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
30X Wash Buffer, 50mL
Absorbance at 450nm minus absorbance at 550nm
<7pg/mL
50┬ÁL
96 microplate wells
Natural and recombinant mouse interleukin-6 (IL-6)
Mouse IL-6 ELISA Kit
Complete ELISA kit with coated 96-well plate
Intra-assay CV <10%; inter-assay CV <10%
1 plate kit
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with mouse GM-CSF, IFN-g, IL-1aa, IL-2, IL-3, IL-4, IL-5, IL-7, IL-9, IL-10, IL-12, or human IL-6. This kit has been shown to have a high level of cross-reactivity with recombinant rat IL-6
4 hr.
Videos