missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-5 Quantikine ELISA Kit, R&D Systems™

Mouse IL-5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SM5000

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452806

  • £2266.74 / Pack of 6


Estimated Shipment date
from Supplier 27-03-2020
Add to basket

Description

Description

The Mouse IL-5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-5 in cell culture supernates (50 ul), serum (50 ul).
Specifications

Specifications

15.60 - 1000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA
7pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Mouse
HRP
Six 96-well plates
16191
Cell Culture Supernates (50uL),Serum (50uL)
1 Pack
Serum
Videos
Documents