missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-5 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure mouse IL-5 in cell culture supernates and mouse serum.

Brand:  RD Systems (Bio-Techne) M5000

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13488215

  • £660.00 / 192 tests


Estimated Shipment
08-03-2018
Add to basket

Description

Description

It contains Sf 21-expressed recombinant mouse IL-5 and antibodies raised against the recombinant factor. This immunoassay has been shown to accurately quantitate recombinant mouse IL-5. Results obtained using natural mouse IL-5 showed linear curves that were parallel to the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural mouse IL-5
Specifications

Specifications

HRP
two 96-well plates
16191
Mouse
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA
1 Kit
Videos