missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse GM-CSF, Recombinant Protein, Carrier-Free

Mouse Recombinant Protein

Brand:  Invitrogen™ 34-8331-85

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15548786

  • £1189.84 / 500µg


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Description

Description

Mouse Granulocyte/Macrophage Colony Stimulating Factor (GM-CSF) is an ∽14 kDa factor produced mainly by activated T cells and macrophages. Other cell types, such as endothelium and fibroblasts, also secrete GM-CSF in response to TNF alpha, IL-2, IL-1, and IFN gamma. GM-CSF stimulates growth of macrophages, granulocytes and dendritic cells. GM-CSF is found as a membrane-bound form and also as a complex associated with the extracellular matrix. Non-glycosylated GM-CSF is biologically active.
Specifications

Specifications

NM_009969
Cytokine Bioassays
14kDa
SDS-PAGE
500μg
Store at less than or equal to -70°C.
E. coli expressed mature mouse GM-CSF accession # NM_009969
Granulocyte/Macrophage-Colony Stimulating Factor
Recombinant Protein, Carrier-Free
Mouse
0.5mg/mL
Sterile liquid; phosphate buffered saline, pH 7.2, 0.22μm filtered.
GM-CSF
>98%
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
The ED50 of this protein, as measured by MC/9 cell proliferation assay, is less than or equal to 650pg/mL. This corresponds to a specific activity of greater than or equal to 1.5 x 106 Units/mg.
Unlabeled
Yes
Videos