missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-4 Quantikine ELISA Kit, R&D Systems™

Mouse IL-4 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SM4000B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442806

  • £2222.64 / Pack of 6

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Mouse IL-4 Quantikine ELISA Kit quantifies mouse IL-4 in cell culture supernates (50 ul), serum (50 ul).
Specifications

Specifications

1 Pack
Mouse
HRP
ELISA
Solid Phase Sandwich ELISA
2pg/mL
Natural and recombinant mouse IL-4
Sufficient materials to run ELISAs on at least six 96-well plates
Serum
7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
16189
6 Plates
Cell Culture Supernates (50uL),Serum (50uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
Documents