missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-4 ELISPOT Ready-SET-Go!™

Contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IL-4-secreting cells. This ELISPOT reagent set is pre-titrated for optimal spot development.

Brand:  Invitrogen™ 88-7844-21

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15501187

  • £152.25 / Pack of 2


Estimated Shipment date
from Supplier 28-03-2018
Add to basket

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISPOT Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 uM).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

Description

Description

The complete, ready-to-use set includes all the reagents needed to perform ELISPOT assays, including: capture and detection antibody, coating buffer powder, assay diluent and detection enzyme. It is supplied with a certificate of anaylsis.

Specifications

Specifications

ELISPOT
Mouse
IL-4
2 x 96 tests
2° to 8°C
Interleukin-4
Videos