missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-28A Mouse ELISA Kit
For the quantitative detection of mouse IL-28.
£387.00 - £1760.00
Specifications
For Use With (Application) | ELISA |
---|---|
Sample Size | 50 μL |
Synonym | IFNL2, IFNL3 Interleukin 28, IL-28A, IL-28B |
Target | Mouse |
Conjugate | Biotin |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-28
Biotin-Conjugate anti-mouse IL-28 monoclonal antibody
Streptavidin-HRP
Mouse IL-28 Standard lyophilized, 4000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
ELISA | |
IFNL2, IFNL3 Interleukin 28, IL-28A, IL-28B | |
Biotin | |
12.85 pg/mL |
50 μL | |
Mouse | |
Supernatant |
For Research Use Only