missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse LIF, Recombinant Protein, Carrier-Free

Mouse Recombinant Protein

Brand:  Invitrogen™ 34-8521-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15598806

  • £411.84 / 100µg


Estimated Shipment date
from Supplier 29-03-2018
Add to basket

Description

Description

Leukemia inhibitory factor (LIF) is a 20 kDa protein that belongs to the IL-6 receptor family. It binds to a heterodimeric membrane receptor made up of a LIF-specific subunit, gp190 or LIFR, and the subunit gp130, which is shared with the other members of the IL-6 family. LIF expression has been observed in various tissues including thymus, lung, and neuronal tissue. LIF can be up-regulated by pro-inflammatory cytokines such as TNFα and IL-17, and elevated levels of LIF have been found in cases of rheumatoid arthritis, neural injury, systemic inflammation, and tuberculosis. LIF displays diverse biological effects, but is best known for its ability to inhibit the differentiation of embryonic stem cells in mice and contribute to stem cell self-renewal.Human and mouse LIF share 79% sequence homology and exhibit cross-species activity. However, LIF inhibition of stem cell differentiation appears to be mouse-specific.
Specifications

Specifications

NM_008501
Cytokine Bioassays
20kDa
SDS-PAGE
100μg
Store at less than or equal to -70°C.
E. coli derived Ser24-Phe203 accession number NM_008501
leukocyte inhibitory factor, Leukemia inhibitory factor
Recombinant Protein, Carrier-Free
Mouse
0.5mg/mL
Sterile liquid: phosphate buffered saline, 0.05% Tween-20, 0.22μm filtered
LIF
>98%
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay
Recombinant mouse LIF has been tested for induction of mIL-6 in the M1 cell line. The ED50 in this assay is 0.75-0.4ng/mL, which corresponds to a specific activity of 1.3 x 106 - 2.5 x 106 Units/mg. When used at 10ng/mL, this recombinant protein has also been found to support mouse embryonic stem cells in an undifferentiated state at same concentration.
Unlabeled
Yes
Videos