missing translation for 'onlineSavingsMsg'
Learn More

Human Activin A, Recombinant Protein, Invitrogen™

Human Recombinant Protein

Brand:  Affymetrix eBioscience 14-8993-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15597556

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Activin A is a member of the TGF-β superfamily, along with the other activin proteins and the structurally related inhibins. The activins are dimeric proteins formed by the association of two of the four existing β subunits, βA, βB, βC, and βE. These subunits are shared with the inhibin proteins, which are heterodimers of one β and one α chain. Activin A is a homodimer of two 13 kDa βA subunits, which can also associate with βB or βC to form Activin AB or AC, respectively. Activin A is expressed by various cell types and exhibits pleiotropic effects including the regulation of metabolism, homeostasis, cell proliferation and differentiation, apoptosis, and tissue healing. Activin A has been recently reported to work in synergy with TGF-β to promote the induction of antigen specific iTreg cells leading to inhibition of Th1 and Th2-mediated responses.
Specifications

Specifications

NM_002192
Cytokine Bioassays
26kDa
SDS-PAGE
25μg
Store at less than or equal to -70°C.
Insect expressed amino acids Gly311-Ser426, accession number NM_002192
The ED50 of this protein, as measured by inhibition of MPC-11 mouse plasmacytoma cell proliferation, is 0.75-3ng/mL. This corresponds to a specific activity of 1.3 x 106 - 3.3 x 105 Units/mg.
Unlabeled
Yes
0.1mg/mL
Sterile liquid; 20mM glycine, 150mM NaCl with 1% BSA, pH 3.0. 0.22μm filtered.
Activin A
≥95%
Insect Cells
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
Recombinant Protein
Human
Videos
Documents