missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-2 ELISPOT Ready-SET-Go!™

Contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IL-2-secreting cells. eBioscience™ Mouse IL-2 ELISPOT Ready-SET-Go!™ reagent set is pre-titrated for optimal spot development.

Brand:  Invitrogen™ 88-7824-21

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15571177

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer Powder.Reconstitute to 1L with dH20 and filter (0.22 uM)).
5X ELISA/ELISPOT Diluent.
Enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

Description

Description

The complete, ready-to-use set includes all the reagents needed to perform ELISPOT assays, including: capture and detection antibody, coating buffer powder, assay diluent and detection enzyme. It is supplied with a certificate of anaylsis.

Specifications

Specifications

2 x 96 Tests
ELISPOT
IL-2
Mouse
2°C to 8°C
Interleukin-2
Videos
Documents

For Research Use Only