missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-2 ELISPOT Ready-SET-Go!™

Contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IL-2-secreting cells. This ELISPOT reagent set is pre-titrated for optimal spot development.

Brand:  Invitrogen™ 88-7824-88

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581177

  • £363.79 / Pack of 10


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer Powder.Reconstitute to 1L with dH20 and filter (0.22 uM)).
5X ELISA/ELISPOT Diluent.
Enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

Description

Description

The complete, ready-to-use set includes all the reagents needed to perform ELISPOT assays, including: capture and detection antibody, coating buffer powder, assay diluent and detection enzyme. It is supplied with a certificate of anaylsis.

Specifications

Specifications

ELISPOT
Mouse
IL-2
10 x 96 tests
2° to 8°C
Interleukin-2
Videos

For Research Use Only