missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-17F (homodimer) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of mouse IL-17FF homodimer protein levels from samples including serum, and cell culture supernatants.

Brand:  Invitrogen™ 88-7472-88

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15531157

  • £280.64 / Pack of 10


Estimated Shipment date
from Supplier 24-07-2018
Add to basket

Alert:

Contains Azide


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

Mouse IL-17F ELISA Ready-SET-Go! Reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
  • The assay demonstrates parallelism in measuring recombinant and native mouse IL-17F proteins and a standard curve range of 15 to 2000pg/mL
  • No crossreactivity to the IL-17AA homodimer is observed and <2.5% crossreactivity to IL-17AF heterodimer when spiked in excess (100ng/ml)
Specifications

Specifications

ELISA
15pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Interleukin-17F, IL-17FF
10 x 96 tests
Mouse
IL-17F
Videos

For Research Use Only