missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-17AF Platinum ELISA Kit

For the quantitative detection of mouse IL-17AF.

Brand:  Invitrogen™ BMS6026

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15551997

  • £352.46 / 96 tests


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-17AF
Biotin-Conjugate anti-mouse IL-17F polyclonal antibody
Streptavidin-HRP
Mouse IL-17AF Standard lyophilized, 500pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (EDTA)
Mouse
Interleukin 17AF, IL-17 A/F heterodimer
ELISA
3.3pg/mL
50μL
IL-17AF
Videos

For Research Use Only