missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17AF (heterodimer) Mouse Uncoated ELISA Kit

Specifically engineered for accurate and precise measurement protein levels from samples including serum and cell culture supernatants.

Brand:  Invitrogen™ 88-8711-76

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15591227

  • £460.00 / Pack of 20


Estimated Shipment date
from Supplier 19-05-2020
Add to basket

Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer 10X PBS ELISA Coating Buffer.
Assay Diluent. 5X concentrated
Sample Diluent A
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

  • Mouse IL-17AF (heterodimer) Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
  • By incorporating a monoclonal antibody to IL-17A as the capture reagent and IL-17AF as the detection reagent, this second generation kit has increased sensitivity with a range from 16 to 2000pg/mL and no cross-reactivity to the IL-17A or IL-17F homodimers
Specifications

Specifications

ELISA
2°C to 8°C except standard which should be stored at ≤-70°C
Interleukin-17A/F
Mouse
16pg/mL
IL-17AF
20 x 96 Tests
Videos
Documents

For Research Use Only