Learn More
Invitrogen™ IL-17AF (heterodimer) Mouse Uncoated ELISA Kit
Specifically engineered for accurate and precise measurement protein levels from samples including serum and cell culture supernatants.
Brand: Invitrogen™ 88-8711-77
349.35 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer 10X PBS ELISA Coating Buffer.
Assay Diluent. 5X concentrated
Sample Diluent A
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution
Description
- Mouse IL-17AF (heterodimer) Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
- By incorporating a monoclonal antibody to IL-17A as the capture reagent and IL-17AF as the detection reagent, this second generation kit has increased sensitivity with a range from 16 to 2000pg/mL and no cross-reactivity to the IL-17A or IL-17F homodimers
Specifications
16pg/mL | |
ELISA | |
20 x 96 Tests | |
Interleukin-17A/F |
Mouse | |
IL-17AF | |
2°C to 8°C except standard which should be stored at ≤-70°C | |
IL-17AF |
For Research Use Only