missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-15/IL-15R Complex Mouse Uncoated ELISA Kit

Uncoated ELISA Kit

Brand:  Invitrogen™ 88-7215-88

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15561077

  • £301.00 / Pack of 10


Estimated Shipment
12-08-2020
Add to basket

Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilutions of antibodies and standards

Description

Description

Interleukin-15 (IL-15) is a 14 kDa proinflammatory protein that has been shown to play a role in the activation of neutrophils, dendritic cells, and macrophages, and is essential to the development and survival of NK cells and CD8 T-cells. IL-15 activates Jak/Stat pathways by signaling through a heterotrimeric receptor. Two of the subunits of this receptor, beta and gamma, are shared with the IL-2 receptor, while the alpha subunit is unique to IL-15.Despite the expression of IL-15 mRNA in many cell types, the secreted protein is rarely detectable in biological samples. Recent research suggests that IL-15 is retained inside the cell and is only secreted in complex with its unique receptor, IL-15R alpha. This chaperoning is required from the ER to Golgi through to secretion. The cytokine/receptor complex may stay bound to the cell surface, where it can be trans-presented to cells expressing the beta/gamma subunits of the receptor, or may be secreted in a soluble form that lacks the transmembrane domain of the receptor. The soluble IL-15/IL-15R complex is produced by mouse dendritic cells, fibroblasts, and macrophages, and demonstrates a 10-100-fold increase in agonistic activity over IL-15 alone.
Specifications

Specifications

P48346,Q60819
HRP
Biotin
16168, 16169
ELISA Kit
4 pg/mL
2°C to 8°C
24 hr. 30 min.
AV080368,Mcp-1
10 x 96 Tests
Supernatant
Colorimetric Microplate Reader
4-500 pg/mL
ELISA
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for the dilutions of antibodies and standards, 96-Well Plates: Corning Costar Flat-bottom (included with Cat. Nos. ending in -22 and -86)
RUO
Serum, 100 μL|Supernatant, 100 μL
1 hr. 45 min.
Mouse
IL-15 Receptor complex with IL-15
Mouse
Videos
Documents

For Research Use Only