Learn More
Invitrogen™ IL-13 Mouse ELISA Kit
For the quantitative detection of mouse IL-13.
Brand: Invitrogen™ BMS6015FIVE
962.71 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Capture Antibody Concentrate (250 x)
Detection Antibody Concentrate (250 x)
mouse IL-19 standard, (lyophilized), 8000pg/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS) 10x
Assay Buffer A Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Z
Tetramethylbenzidine (TMB) Substrate Solution
2 96-well plates
Description
The Mouse Interleukin-13 (Ms IL-13) ELISA quantitates Ms IL-13 in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-13. Principle of the method The Mouse IL-13 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
2.8pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum | |
ELISA | |
50 μL | |
IL-13 |
Biotin | |
Mouse | |
5 x 96 Tests | |
ALRH, BHR1, allergic rhinitis |
For Research Use Only