missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-12, Recombinant Protein

Mouse Recombinant Protein

Brand:  Invitrogen™ 14-8121-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15587496

  • £368.01 / 25µg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Interleukin-12 (IL-12) is a heterodimeric 70 kD (p70) cytokine composed of two covalently linked, glycosylated chains, 40kD (p40) and 35-kD (p35). IL-12 is mainly produced by monocytes, macrophages, and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS), to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma (IFN-g) production, cell proliferation, and cytotoxicity mediated by natural killer cells and T cells. It is now established that IL-12 also plays a key role in the development of Th1 responses, leading to IFN-g and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation.

eBioscience’s recombinant mouse IL-12 p70 is produced in baculovirus-infected insect cells as an authentic heterodimer of precursor p35 and p40 subunits using a dual promoter expression system. It is distinct from other available forms of the protein in that it is expressed as a true heterodimer, as opposed to a single-chain, pseudo-heterodimer in which the subunits are joined by an artificial linker.
Specifications

Specifications

NM_008352 co-expressed with NM_008351
Sterile liquid; 10mM NaH2PO4, pH 6.0, 300mM NaCl, with 1.0% BSA. 0.22μm filtered.
IL-12
>98%
Insect cells infected with baculovirus
RUO
Less than 0.01ng/μg cytokine, as determined by the LAL assay.
The ED50 of this protein, as measured by IFN gamma induction assay in mouse splenocytes, is less than or equal to 175pg/mL. This corresponds to a specific activity of greater than or equal to 5.7 x 106 Units/mg.
Unlabeled
Yes
Cytokine Bioassays , ELISA Recombinant Standard
The heterodimeric, p40, amino acids met 23-ser 335, cystine linked to p35, amino acids arg 23-ala 215, has a predicted molecular mass of 57,496. On non-reducing SDS-PAGE the heterodimeric cystine-linked protein migrates as a 65kDa protein. The DTT reduced protein migrates as 43kDa and 24kDa polypeptides.
SDS-PAGE
25μg
Store at less than or equal to -70°C.
Insect cells infected with baculovirus: mouse p40, amino acids met 1-ser 335, accession # NM_008352 was co-expressed with mouse p35, amino acids met 1-ala 215, accession # NM_008351
Interleukin-12, IL12, p70
Recombinant Protein
Mouse
Videos

For Research Use Only.