missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-12 Mouse ELISA Kit
For the quantitative detection of mouse IL-12.
£451.00 - £2400.00
Specifications
Accession Number | P43431,P43432 |
---|---|
Conjugate | Biotin |
Product Type | ELISA Kit |
Sample Type | Serum, Supernatant |
Target Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15542047
|
Invitrogen™
BMS616TEN |
10 x 96 Tests |
£2400.00
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15532047
|
Invitrogen™
BMS616 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
The Mouse Interleukin-12 (Ms IL-12) ELISA quantitates Ms IL-12 in mouse serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-12. Principle of the method The Mouse IL-12 solid-phase sandwich ELI
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
P43431,P43432 | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
5% | |
RUO | |
2°C to 8°C | |
Mouse | |
3 hr. 10 min. |
Biotin | |
Serum, Supernatant | |
ELISA | |
10% | |
HRP | |
Serum, 50 μL; Supernatant, 50 μL | |
CMLF, NKSF, Interleukin 12 | |
1 hr. 20 min. |
For Research Use Only