missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12 p70 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure mouse IL-12 p70 in cell culture supernates and mouse serum.

Brand:  R&D Systems™ M1270

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13458205

  • £645.09 / 192 tests


Estimated Shipment
04-03-2019
Add to basket

Description

Description

It contains recombinant mouse IL-12 expressed in Sf 21 insect cells using a baculovirus expression vector and antibodies raised against recombinant mouse IL-12. This immunoassay has been shown to accurately quantitate the recombinant factor. Results obtained using natural mouse IL-12 showed linear curves that were parallel to the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural mouse IL-12 p70
Specifications

Specifications

7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16159
2 Plates
2.5pg/mL
Cell Culture Supernates (50μL), Serum (50μL)
Natural and recombinant mouse IL-12 p70
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL), Serum (50uL)
Mouse
Sufficient materials to run ELISAs on at least two 96-well plates
Videos