missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-12 (p70) ELISA Kits

Measure levels of active mouse interleukin-12, p70 in mouse serum and culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ EMIL12

Code : PN

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10648084

  • £405.75 / 1 set


Estimated Shipment
03-07-2018
Add to basket

Description

Description

These mouse IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of biologically active interleukin-12 (IL-12 p70) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Biologically active (p70) natural and recombinant mouse interleukin-12 (IL-12, p70)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96 well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 12.3 to 1000pg/mL

  • Sensitivity: < 5pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-1α, IL-1β, IL-3, IL-4, IL-5, IL-6, IL-7, TNFα, IFNγ, GM-CSF or human IL-12 (p70). Recombinant mouse IL-12 (p40) and human IL-12 < (p40) do not cross-react or interfere with this assay

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Specifications

Specifications

12.3 to 1000pg/mL
Colorimetric ELISA (TMB substrate) in clear 96 well strip plate
30X Wash Buffer, 50mL, Biotinylated Antibody Concentrate, 125µL, Streptavidin-HRP Concentrate, 75µL, Stop Solution, 13mL, Standard Diluent, 12mL, Lyophilized Recombinant Mouse IL-12 p70 Standard, 2 vials, HRP Dilution Buffer, 14mL, Biotinylated Antibody Diluent, 10mL, Anti-Mouse IL-12 p70 Precoated 96-well Strip Plate, 1 ea, Adhesive Plate Covers, 6 ea, TMB Substrate Solution, 13mL
Absorbance at 450nm minus absorbance at 550nm
<5pg/mL
50µL
96 microplate wells
Biologically active (p70) natural and recombinant mouse interleukin-12 (IL-12, p70)
Mouse IL-12 (p70) ELISA Kit
Complete ELISA kit with coated 96-well plate
Intra-assay CV <10%; inter-assay CV <10%
1 plate kit
Cell culture supernatant, serum
No significant cross-reaction with mouse IL-1α, IL-1β, IL-3, IL-4, IL-5, IL-6, IL-7, TNFα, IFNγ, GM-CSF or human IL-12 (p70). Recombinant mouse IL-12 (p40) and human IL-12 < (p40) do not cross-react or interfere with this assay
3 hr.
Videos
Documentation

Technical Documents