Thermo Scientific™ Pierce™ Mouse IL-12 (p70) ELISA Kits

Measure levels of active mouse interleukin-12, p70 in mouse serum and culture supernatant samples using 96-well plates.

Overview
Brand: Thermo Scientific™ Pierce™

Manufacturer Part Number: EMIL122

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10391104

Quantity Price
1 £ 524.0 / 1 set
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Specificity No significant cross-reaction with mouse IL-1α, IL-1β, IL-3, IL-4, IL-5, IL-6, IL-7, TNFα, IFNγ, GM-CSF or human IL-12 (p70). Recombinant mouse IL-12 (p40) and human IL-12 < (p40) do not cross-react or interfere with this assay
Sensitivity <5pg/mL
Precision Intra-assay CV <10%; inter-assay CV <10%
Measurement Absorbance at 450nm minus absorbance at 550nm
Formulation Complete ELISA kit with coated 96-well plates
Kit Contents 30X Wash Buffer, 2 x 50mL
Assay Range 12.3 to 1000pg/mL
Sufficient For 2 x 96 microplate wells
Format Colorimetric ELISA (TMB substrate) in clear 96 well strip plate
Description Mouse IL-12 (p70) ELISA Kit
Sample Type Cell culture supernatant, serum
Sample Size 50µL
Target Biologically active (p70) natural and recombinant mouse interleukin-12 (IL-12, p70)
Quantity 2 plate kit
Total Assay Time 3 hr.

These mouse IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of biologically active interleukin-12 (IL-12 p70) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Biologically active (p70) natural and recombinant mouse interleukin-12 (IL-12, p70)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96 well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 12.3 to 1000pg/mL

  • Sensitivity: < 5pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-1α, IL-1β, IL-3, IL-4, IL-5, IL-6, IL-7, TNFα, IFNγ, GM-CSF or human IL-12 (p70). Recombinant mouse IL-12 (p40) and human IL-12 < (p40) do not cross-react or interfere with this assay

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Documentation

Technical documents