missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet, R&D Systems™

Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DY2398

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13433345

  • £490.98 / Pack of 15


Estimated Shipment date
from Supplier 01-04-2020
Add to basket

Description

Description

The Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-12/IL-23 p40.
Specifications

Specifications

ELISA
Biotin
16160
15 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
Mouse
15.60 to 1000 pg/mL
Videos
Documents