missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12/IL-23 p40 Allele-specific Quantikine ELISA Kit, R&D Systems™

Mouse IL-12 p40 Allele-specific Quantikine ELISA Kt from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ M1240

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13448205

  • £783.51 / 192 tests

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Mouse IL-12 p40 Allele-specific Quantikine ELISA Kt is a Solid Phase Sandwich ELISA that quantifies mouse IL-12/IL-23 p40 in cell culture supernates (50 ul), serum (50 ul).
Specifications

Specifications

11.70 - 750pg/mL
<50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules
16160
1 Kit
Cell Culture Supernates (50uL), Serum (50uL)
Mouse
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
2 Plates
4pg/mL
Cell Culture Supernates (50μL), Serum (50μL)
Natural and recombinant mouse IL-12/IL-23 p40
Sufficient materials to run ELISAs on at least two 96-well plates
Videos
Documents