missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-10 Mouse ELISA Kit

For the quantitative detection of mouse IL-10.

£352.46 - £1570.00

Specifications

For Use With (Application) ELISA
Sample Size 50 μL
Synonym TGIF, MCSF III, CSIF, B TCGF
Target IL-10
Conjugate Biotin
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-10
Biotin-Conjugate anti-mouse IL-10 polyclonal antibody
Streptavidin-HRP
Mouse IL-10 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15592037 View Documents Invitrogen™
BMS614-2FIVE
5 x 96 Tests £1035.00
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Add to basket
 
15502047 View Documents Invitrogen™
BMS614-2TEN
10 x 96 Tests £1570.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Add to basket
 
15582037 View Documents Invitrogen™
BMS614-2
96 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
15512047 View Documents Invitrogen™
BMS614-2TWO
2 x 96 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

ELISA
TGIF, MCSF III, CSIF, B TCGF
Biotin
5.0pg/mL
50 μL
IL-10
Serum

For Research Use Only