missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-10 Mouse ELISA Kit

For the quantitative detection of mouse IL-10.

Brand:  Invitrogen™ BMS614-2FIVE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15592037

  • £1035.00 / Pack of 5


Estimated Shipment date
from Supplier 20-04-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-10
Biotin-Conjugate anti-mouse IL-10 polyclonal antibody
Streptavidin-HRP
Mouse IL-10 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

Biotin
5.0pg/mL
IL-10
5 x 96 Tests
Serum
ELISA
50 μL
TGIF, MCSF III, CSIF, B TCGF
Mouse
Videos
Documents

For Research Use Only