missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IL-10 Platinum ELISA Kit

For the quantitative detection of mouse IL-10.

Brand:  Invitrogen™ BMS614-2TWO

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15512047

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-10
Biotin-Conjugate anti-mouse IL-10 polyclonal antibody
Streptavidin-HRP
Mouse IL-10 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

Biotin
2 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
Mouse
TGIF, MCSF III, CSIF, B TCGF
ELISA
5.0pg/mL
50μL
IL-10
Videos

For Research Use Only