Learn More
Invitrogen™ IL-10 Mouse Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of mouse IL-10 protein levels from samples including serum and cell culture supernatants. This 2nd Generation kit has decreased background and improved performance in serum samples.
£173.00 - £546.00
Specifications
Accession Number | P18893 |
---|---|
Assay Range | 32-4,000 pg/mL |
Assay Sensitivity | 32 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15501047
|
Invitrogen™
88-7105-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15571037
|
Invitrogen™
88-7105-22 |
2 x 96 Tests |
£173.00
Pack of 2 |
In Stock
Log in to see stock. |
|||||
15581037
|
Invitrogen™
88-7105-76 |
20 x 96 Tests |
£546.00
Pack of 20 |
In Stock
Log in to see stock. |
|||||
Description
The Mouse Interleukin 10 (IL-10) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-10. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Mouse IL-10 Ready-SET-Go! ELISA (2nd Generation) Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
P18893 | |
32 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
IL-23,p19 | |
HRP | |
Serum, 100 μL; Supernatant, 100 μL | |
Interleukin 10, CSIF, Cytokine Synthesis Inhibitory Factor | |
1 hr. 45 min. |
32-4,000 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
16153 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, Coating Buffer 10X PBS ELISA Coating Buffer., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86) | |
RUO | |
2°C to 8°C | |
Mouse | |
24 hr. 30 min. |
For Research Use Only