Thermo Scientific Mouse IL-10 ELISA Kits

Measure levels of mouse interleukin-10 in mouse serum and cell culture supernatant samples using 96-well plates.

Overview
Brand: Thermo Scientific

Manufacturer Part Number: EM2IL105

MOUSE IL-10 ELISA

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10741175

Quantity Price
1 £ 1360.0 / 1 set
Estimated Shipment date
from Supplier 08-03-2017
Add to basket


Description and Specification

Specification

Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <12pg/mL
Description Mouse IL-10 ELISA Kit
Sample Size 50µL
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Formulation Complete ELISA kit with coated 96-well plates
Specificity No significant cross-reaction with mouse IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, GM-CSF, IFNg, TNFa or human IL-10
Target Natural and recombinant mouse interleukin-10 (IL-10)
Total Assay Time 5 hr.
Kit Contents 30X Wash Buffer, 200mL
Assay Range 37 to 3000pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 5 plate kit
Sample Type Cell culture supernatant, serum
Sufficient For 5 x 96 microplate wells

These mouse IL-10 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-10 (IL-10) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse interleukin-10 (IL-10)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 37 to 3000pg/mL

  • Sensitivity: < 12pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, GM-CSF, IFNγ, TNFα or human IL-10

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Documentation

Technical documents