missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1 beta Quantikine ELISA Kit, R&D Systems™

Mouse IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ MLB00C

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462176

  • £755.58 / 192 tests


Estimated Shipment date
from Supplier 24-03-2020
Add to basket

Description

Description

The Mouse IL-1 beta/IL-1F2 Quantikine ELISA Kit quantifies mouse IL-1 beta/IL-1F2 in cell culture supernates (50 ul), tissue lysates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications

Specifications

1 Kit
Mouse
HRP
ELISA
Solid Phase Sandwich ELISA
4.8pg/mL
Natural and recombinant mouse IL-1 beta
Sufficient materials to run ELISAs on at least two 96-well plates
Tissue Lysates
12.50 - 800pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16176
2 Plates
Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
Documents