missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-1 beta/IL-1F2 Quantikine ELISA Kit
Mouse IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 12.50 - 800pg/mL |
---|---|
Assay Sensitivity | 4.8pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates |
Description
The Mouse IL-1 beta/IL-1F2 Quantikine ELISA Kit quantifies mouse IL-1 beta/IL-1F2 in cell culture supernates (50 ul), tissue lysates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).Specifications
12.50 - 800pg/mL | |
4.8pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates |