missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgM Mouse Uncoated ELISA Kit with Plates

For quantitative detection of mouse IgM.

Brand:  Invitrogen™ 88-50470-86

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15590907

  • £322.00 / Pack of 10


In stock, ready to ship: 2
Add to basket

Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
mouse IgM isotype control (standard), lyophilized, 50ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Tetramethylbenzidine (TMB) Substrate Solution
96 Well Plate included with product Cat. #’s ending in suffixes -22, -76, -86

Description

Description

Immunoglobulin M is the third most common serum Ig and can exist as both a pentamer and as a monomer (e.g. found on B lymphocytes as B - cell receptors). The large pentameric structure allows for building of bridges between encountered epitopes on molecules that are too distant as to be connected by smaller IgG antibodies. IgM is built as the first antibody during an immune response and is responsible for agglutination and cytolytic reactions since ist pentameric structure gives it 10 free antigen binding sites in theory and it posseses a high avidity. Due to conformational constraints among the 10 Fab portions, IgM only has a valence of 5. Additionally, IgM is not as versatile as IgG, but it is of vital importance in complement activation and agglutination. IgM is predominantly found in the lymph fluid and the blood and is a very effective neutralizing agent in the early stages of disease. Elevated levels can be a sign of recent infection or exposure to antigen.
Specifications

Specifications

0.39-25 ng/mL
ELISA
ELISA Kit
0.39 ng/mL
Plasma, 10 μL|Serum, 10 μL|Supernatant, 50 μL
1 hr. 45 min.
Mouse
10 x 96 Tests
Supernatant
Colorimetric Microplate Reader
HRP
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
RUO
50 μL (prediluted 1:10,000)
2°C to 8°C
24 hr. 30 min.
Immunoglobulin M
Mouse
HRP
Videos
Documents

For Research Use Only