missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IgG2b ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of mouse IgG2b.

Brand:  Invitrogen™ 88-50430-86

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15500897

  • £239.00 / Pack of 10


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Disclaimers:

For Research Use Only


Includes: Capture Antibody Concentrate

Description

Description

Mouse IgG2b Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
10 x 96 tests
Cell culture supernatant, serum and plasma
Mouse
IgG2b
96-well plates
3.90ng/mL
50μL (prediluted 1:10,000)
2° to 8°C
Immunoglobulin G2b
Videos