missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2a Isotype Control (PPV-04), Alexa Fluor™ 488, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA518169
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This product can be used as a negative control to measure the non-specific binding of Mouse IgG2a in human, porcine, mouse and rat tissues.
Mouse IgG2a Control antibody specifically detects Mouse IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Mouse IgG2a | |
PPV-04 | |
Alexa Fluor 488 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Size-exclusion chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a |