missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1, Isotype Control, Invitrogen™

Mouse Isotype Control

Brand:  Invitrogen MA518092

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15397364

  • £233.44 / 100 tests


Estimated Shipment
31-05-2018
Add to basket

Description

Description

MA5-18092 is negative on all human cells and cell lines tested. This antibody recognizes a rat cell surface marker, and therefore cannot be used as a negative control in this species. Test results have shown that MA5-18092 is also suitable for use as a negative control with bovine, ovine, porcine, equine, canine, lapine and guinea-pig tissues.For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
Specifications

Specifications

Mouse IgG1
Control
Unconjugated
Purified
Murine
IgG1
100 Tests
Isotype Control
Immunohistochemistry (Frozen)
0.1mg/mL
PBS with 1% BSA and 0.09% sodium azide
IgG1
Rat thymocyte membrane glycoproteins
Protein A
Store at 4°C or Store at -20°C if preferred
Not Applicable
Videos