Learn More
Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor™ 488, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA518167
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This product can be used as a negative control to measure the non-specific binding of Mouse IgG1 in human and rat tissues; it has been tested on a variety of resting, activated, live and fixed rat and human tissues.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Mouse IgG1 | |
MOPC-21 | |
Alexa Fluor 488 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Size-exclusion chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |